Anti-KRT7 Mouse Monoclonal Antibody [clone: OV-TL12/30]

ABCAAB9098-250EA 640 EUR
ABCAAB9098-250
Anti-KRT7 Mouse Monoclonal Antibody [clone: OV-TL12/30]
Type: Primary
Antigen: KRT7 (keratin 7)
Clonality: monoclonal
Clone: OV-TL12/30
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR