Anti-Rhodamine Mouse Monoclonal Antibody [clone: 11H10]

Proveedor: Abcam
ABCAAB9093-100EA 806 EUR
ABCAAB9093-100
Anti-Rhodamine Mouse Monoclonal Antibody [clone: 11H10]
Anticuerpos
Type: Primary
Antigen: Rhodamine
Clonality: monoclonal
Clone: 11H10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR