Anti-Maltose Binding Protein Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9084-100EA 600 EUR
ABCAAB9084-100
Anti-Maltose Binding Protein Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Maltose Binding Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR