Anti-HTLV II p19 Mouse Monoclonal Antibody [clone: 78/6.18.07]

ABCAAB9075-100EA 0 EUR
ABCAAB9075-100
Anti-HTLV II p19 Mouse Monoclonal Antibody [clone: 78/6.18.07]
Type: Primary
Antigen: HTLV II p19
Clonality: monoclonal
Clone: 78/6.18.07
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR