Anti-HIV1 p17 Mouse Monoclonal Antibody [clone: 2D11]

Proveedor: Abcam
ABCAAB9067-100EA 0 EUR
ABCAAB9067-100
Anti-HIV1 p17 Mouse Monoclonal Antibody [clone: 2D11]
Anticuerpos
Type: Primary
Antigen: HIV1 p17
Clonality: monoclonal
Clone: 2D11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR