Anti-IFIH1 Goat polyclonal antibody

Proveedor: Abcam
ABCAAB4544-100EA 550 EUR
ABCAAB4544-100
Anti-IFIH1 Goat polyclonal antibody
Type: Primary
Antigen: IFIH1 (interferon induced with helicase C domain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR