Anti-DNMT3B Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB2851-100EA 585 EUR
ABCAAB2851-100
Anti-DNMT3B Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: DNMT3B (DNA (cytosine-5-)-methyltransferase 3 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR