Anti-XIAP Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB2541-50EA 555 EUR
ABCAAB2541-50
Anti-XIAP Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: XIAP (X-linked inhibitor of apoptosis)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR