Anti-Keratan Sulfate Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)

Proveedor: Biorbyt
BIRBORB129232-100EA 0 EUR
BIRBORB129232-100
Anti-Keratan Sulfate Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)
Anticuerpos
Type: Primary
Antigen: Keratan Sulfate
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR