Anti-ITGB1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB127588-200EA 0 EUR
BIRBORB127588-200 BIRBORB127588-100
Anti-ITGB1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ITGB1 (integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR