Anti-PRKCQ Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB125368-200EA 0 EUR
BIRBORB125368-200 BIRBORB125368-100
Anti-PRKCQ Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PRKCQ (protein kinase C, theta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR