Anti-FGF23 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB125186-100EA 0 EUR
BIRBORB125186-100
Anti-FGF23 Goat Polyclonal Antibody
Anticuerpos
ELISA: 1:16000

Type: Primary
Antigen: FGF23 (fibroblast growth factor 23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR