Anti-SLCO2B1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Proveedor: Biorbyt
BIRBORB113866-100EA 0 EUR
BIRBORB113866-100
Anti-SLCO2B1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
Type: Primary
Antigen: SLCO2B1 (solute carrier organic anion transporter family, member 2B1)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR