Anti-GDNF Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB66950-100EA 0 EUR
BIRBORB66950-100
Anti-GDNF Rabbit polyclonal antibody
Anticuerpos
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: GDNF (glial cell derived neurotrophic factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR