Anti-PGF Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB66903-100EA 0 EUR
BIRBORB66903-100
Anti-PGF Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: PGF (placental growth factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR