Anti-CCL17 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB66870-100EA 0 EUR
BIRBORB66870-100
Anti-CCL17 Rabbit polyclonal antibody
Anticuerpos
ELISA: 1:1000-1:10000

Type: Primary
Antigen: CCL17 (chemokine (C-C motif) ligand 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR