Anti-EHMT1 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB66842-100EA 0 EUR
BIRBORB66842-100
Anti-EHMT1 Rabbit polyclonal antibody
Anticuerpos
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: EHMT1 (euchromatic histone-lysine N-methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR