Anti-RAC2 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Proveedor: Biorbyt
BIRBORB66477-100EA 0 EUR
BIRBORB66477-100
Anti-RAC2 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Western Blot: 1-1000
Type: Primary
Antigen: RAC2 (ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2))
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat