Anti-GGA2 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB49087-200EA 0 EUR
BIRBORB49087-200 BIRBORB49087-100
Anti-GGA2 Rabbit polyclonal antibody
Anticuerpos
Type: Primary
Antigen: GGA2 (golgi-associated, gamma adaptin ear containing, ARF binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR