Anti-STUB1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB49067-200EA 0 EUR
BIRBORB49067-200 BIRBORB49067-100
Anti-STUB1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: STUB1 (STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR