Anti-CSRP1 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB48990-100EA 0 EUR
BIRBORB48990-100 BIRBORB48990-200
Anti-CSRP1 Rabbit polyclonal antibody
Anticuerpos
Type: Primary
Antigen: CSRP1 (cysteine and glycine-rich protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR