Anti-CEACAM5 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB48942-100EA 0 EUR
BIRBORB48942-100 BIRBORB48942-200
Anti-CEACAM5 Rabbit polyclonal antibody
Anticuerpos
Type: Primary
Antigen: CEACAM5 (carcinoembryonic antigen-related cell adhesion molecule 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR