Anti-MRSA Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB48821-200EA 0 EUR
BIRBORB48821-200
Anti-MRSA Mouse Monoclonal Antibody
Anticuerpos
ELISA: 1:1000-5000; Western Blot: 0.1-0.2 ug/ml

Type: Primary
Antigen: MRSA
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR