Anti-Omentin Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB48720-200EA 0 EUR
BIRBORB48720-200
Anti-Omentin Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Omentin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR