Anti-GHRL Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB48713-200EA 0 EUR
BIRBORB48713-200
Anti-GHRL Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: GHRL (ghrelin/obestatin prepropeptide)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR