Anti-AIMP2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB48686-100EA 0 EUR
BIRBORB48686-100
Anti-AIMP2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: AIMP2 (aminoacyl tRNA synthetase complex-interacting multifunctional protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR