Anti-CT1 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB48655-200EA 0 EUR
BIRBORB48655-200
Anti-CT1 Rabbit polyclonal antibody
Anticuerpos
Type: Primary
Antigen: CT1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR