Anti-S100B Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB48639-200EA 0 EUR
BIRBORB48639-200
Anti-S100B Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: S100B (S100 calcium binding protein B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR