Anti-Neurokinin A Chicken Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB48635-200EA 0 EUR
BIRBORB48635-200
Anti-Neurokinin A Chicken Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Neurokinin A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR