Anti-SCARB1 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB48529-100EA 0 EUR
BIRBORB48529-100
Anti-SCARB1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SCARB1 (scavenger receptor class B, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat