Anti-IL1RN Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB48417-100EA 0 EUR
BIRBORB48417-100
Anti-IL1RN Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: IL1RN (interleukin 1 receptor antagonist)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR