Anti-DKC1 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB48353-100EA 0 EUR
BIRBORB48353-100
Anti-DKC1 Rabbit polyclonal antibody
Anticuerpos
Type: Primary
Antigen: DKC1 (dyskeratosis congenita 1, dyskerin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR