Anti-DCN Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB48349-100EA 0 EUR
BIRBORB48349-100
Anti-DCN Rabbit polyclonal antibody
Anticuerpos
Type: Primary
Antigen: DCN (decorin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR