Anti-CCL1 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB47992-100EA 0 EUR
BIRBORB47992-100
Anti-CCL1 Rabbit polyclonal antibody
Anticuerpos
Type: Primary
Antigen: CCL1 (chemokine (C-C motif) ligand 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR