Anti-IL2RA Rat Monoclonal Antibody (FITC (Fluorescein))

Proveedor: Biorbyt
BIRBORB44838-0.1EA 0 EUR
BIRBORB44838-0.1
Anti-IL2RA Rat Monoclonal Antibody (FITC (Fluorescein))
Anticuerpos
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR