Anti-LCP2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB44777-0.1EA 0 EUR
BIRBORB44777-0.1
Anti-LCP2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: LCP2 (lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of 76kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR