Anti-BSG Mouse monoclonal antibody

Proveedor: Biorbyt
BIRBORB44443-0.1EA 0 EUR
BIRBORB44443-0.1
Anti-BSG Mouse monoclonal antibody
Anticuerpos
Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR