Anti-CD2 Mouse Monoclonal Antibody (Biotin)

Proveedor: Biorbyt
BIRBORB44378-0.1EA 0 EUR
BIRBORB44378-0.1
Anti-CD2 Mouse Monoclonal Antibody (Biotin)
Anticuerpos
Type: Primary
Antigen: CD2 (CD2 molecule)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR