Anti-CD14 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Proveedor: Biorbyt
BIRBORB44085-100EA 0 EUR
BIRBORB44085-100
Anti-CD14 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR