Anti-CD2 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB43932-100EA 0 EUR
BIRBORB43932-100
Anti-CD2 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: CD2 (CD2 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR