Anti-CD38 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB43856-100EA 0 EUR
BIRBORB43856-100
Anti-CD38 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: CD38 (CD38 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR