Anti-PTPRC Rat Monoclonal Antibody (Peridinin Chlorophyll)

Proveedor: Biorbyt
BIRBORB43769-0.1EA 0 EUR
BIRBORB43769-0.1
Anti-PTPRC Rat Monoclonal Antibody (Peridinin Chlorophyll)
Anticuerpos
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone:
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR