Anti-TCF4 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB43571-0.1EA 0 EUR
BIRBORB43571-0.1
Anti-TCF4 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TCF4 (transcription factor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR