Anti-N-Cadherin Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB43408-100EA 0 EUR
BIRBORB43408-100
Anti-N-Cadherin Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: N-Cadherin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR