Anti-CHEK2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB43360-100EA 0 EUR
BIRBORB43360-100
Anti-CHEK2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CHEK2 (checkpoint kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR