Anti-GST tag Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB43336-100EA 0 EUR
BIRBORB43336-100
Anti-GST tag Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: GST tag (Glutathione-S-Transferase tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR