Anti-IgG kappa Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB43303-100EA 0 EUR
BIRBORB43303-100
Anti-IgG kappa Mouse Monoclonal Antibody
Anticuerpos
Type: Secondary
Antigen: Rabbit IgG (kappa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR