Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Biorbyt
BIRBORB43301-100EA 0 EUR
BIRBORB43301-100
Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR