Anti-hnRNP C1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB41799-100EA 0 EUR
BIRBORB41799-100
Anti-hnRNP C1 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:250; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: hnRNP C1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR