Anti-Intestinal alkaline phosphatase Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB41727-100EA 0 EUR
BIRBORB41727-100
Anti-Intestinal alkaline phosphatase Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500

Type: Primary
Antigen: Intestinal alkaline phosphatase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR